accessibility menu, dialog, popup

UserName

CH3HNOONHH3COO

CAS RN 8049-97-6

IUPAC Name: 6,14-dimethyl-4,12-diazapentacyclo[8.6.1.1²,⁵.0¹³,¹⁷.0⁹,¹⁸]octadeca-1(17),2,5,9(18),10,13-hexaene-7,8,15,16-tetrone
Synonyms: melanin melanins
Molecular Weight (g/mol): 318.29
Molecular Formula: C18H10N2O4
InChi Key: XUMBMVFBXHLACL-UHFFFAOYSA-N
SMILES: CC1=C2NC=C3C2=C(C2=CNC4=C(C)C(=O)C(=O)C3=C24)C(=O)C1=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results