CAS RN 80288-49-9
CAS RN 80288-49-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Furafylline, TRC
CAS: 80288-49-9 Molecular Formula: C12 H12 N4 O3 Molecular Weight (g/mol): 260.25 Synonym: 3-(2-furanylmethyl)-3,9-dihydro-1,8-dimethyl-1H-purine-2,6-dione IUPAC Name: 3-(furan-2-ylmethyl)-1,8-dimethyl-7H-purine-2,6-dione SMILES: CN1C(=O)N(Cc2occc2)c3nc(C)[nH]c3C1=O
2
Furafylline, MedChemExpress
MedChemExpress Furafylline is a potent and selective inhibitor of human cytochrome P450IA2 with an IC50 of 0.07 μM.