missing translation for 'onlineSavingsMsg'
Learn More

Furafylline, MedChemExpress

Product Code. 16411079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411079 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411079 Supplier Medchem Express Supplier No. HY107204
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Furafylline is a potent and selective inhibitor of human cytochrome P450IA2 with an IC50 of 0.07 μM.

Furafylline is a potent and selective inhibitor of human cytochrome P450IA2 with an IC50 of 0.07 μM.

Specifications

Chemical Name or Material Furafylline
Physical Form Solid
CAS 80288-49-9
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C12H12N4O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 12.5 mg/mL (48.03 mM; Need ultrasonic)
SMILES O=C1N(C2=C(N=C(C)N2)C(N1C)=O)CC3=CC=CO3
Molecular Weight (g/mol) 260.25
Formula Weight 260.25
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.