accessibility menu, dialog, popup

UserName

H3CNClSOOO

CAS RN 80-77-3

IUPAC Name: 2-(4-chlorophenyl)-3-methyl-1λ⁶,3-thiazinane-1,1,4-trione
Synonyms: chlormezanone tanafol rilassol (+-)-fenarol 氯美扎酮 miorilax banabin chlormezanone preparation banabin-syntyal chlormezanone (inn)
Molecular Weight (g/mol): 273.73
Molecular Formula: C11H12ClNO3S
InChi Key: WEQAYVWKMWHEJO-UHFFFAOYNA-N
SMILES: CN1C(C2=CC=C(Cl)C=C2)S(=O)(=O)CCC1=O
Quantity 
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results