CAS RN 79146-60-4
CAS RN 79146-60-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
N-[4-(Methoxymethyl)-1-[2-(3-thienyl)ethyl]-4-piperidinyl]-N-phenyl-propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[4-(Methoxymethyl)-1-[2-(3-thienyl)ethyl]-4-piperidinyl]-N-phenyl-propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-[4-(Methoxymethyl)-1-[2-(3-thienyl)ethyl]-4-piperidinyl]-N-phenyl-propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.