accessibility menu, dialog, popup

UserName

CAS RN 79055-67-7

NH3(S)POOOOO

CAS RN 79055-67-7

IUPAC Name: (2S)-2-azaniumyl-5-phosphonatopentanoate
Synonyms: (2S)-2-ammonio-5-phosphonatopentanoate
Molecular Weight (g/mol): 195.11
Molecular Formula: C5H10NO5P
InChi Key: VOROEQBFPPIACJ-BYPYZUCNSA-L
SMILES: [NH3+][C@@H](CCCP([O-])([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results