accessibility menu, dialog, popup

UserName

CAS RN 78957-85-4

ClSOONHH2NHCl

CAS RN 78957-85-4

IUPAC Name: N-(2-aminoethyl)-5-chloronaphthalene-1-sulfonamide hydrochloride
Synonyms: N-(2-aminoethyl)-5-chloronaphthalene-1-sulfonamide hydrochloride
Molecular Weight (g/mol): 321.22
Molecular Formula: C12H14Cl2N2O2S
InChi Key: VWAGIWCLJAQLAL-UHFFFAOYSA-N
SMILES: Cl.NCCNS(=O)(=O)C1=C2C=CC=C(Cl)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results