CAS RN 77086-22-7
CAS RN 77086-22-7
IUPAC Name:
(1S,9R)-1-methyl-16-azatetracyclo[7.6.1.0²,⁷.0¹⁰,¹⁵]hexadeca-2,4,6,10,12,14-hexaene; (2Z)-but-2-enedioic acid
Synonyms:
dizocilpine
Molecular Weight (g/mol):
337.38
Molecular Formula:
C20H19NO4
InChi Key:
QLTXKCWMEZIHBJ-PJGJYSAQSA-N
SMILES:
OC(=O)\C=C/C(O)=O.C[C@]12N[C@H](CC3=CC=CC=C13)C1=CC=CC=C21
1
–
3
of
3
results
1
Dizocilpine maleate, 99+%
CAS: 77086-22-7 Molecular Formula: C20H19NO4 Molecular Weight (g/mol): 337.375 MDL Number: MFCD00082465 InChI Key: QLTXKCWMEZIHBJ-PJGJYSAQSA-N Synonym: dizocilpine maleate,+-mk-801 hydrogen maleate,+-mk 801 maleate,dizocilpine hydrogen maleate,unii-6lr8c1b66q,dizocilpine maleate usan,dsstox_cid_25785,dsstox_rid_81126 PubChem CID: 6420042 SMILES: CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O
2
Tocris Bioscience™ (+)-MK 801 maleate
CAS: 77086-22-7 Molecular Formula: C20H19NO4
3
Dizocilpine maleate, MedChemExpress
MedChemExpress Dizocilpine maleate (MK-801 maleate) is a potent, selective and non-competitive NMDA receptor antagonist with Kd of 37.2 nM in rat brain membranes.