accessibility menu, dialog, popup

UserName

H3C(S)OH(R)OO(R)(S)OHH2C(S)CH2(S)(R)HOO

CAS RN 77-06-5

IUPAC Name: (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Synonyms: gibberellic acid
Molecular Weight (g/mol): 346.38
Molecular Formula: C19H22O6
InChi Key: IXORZMNAPKEEDV-QTWFBFKQSA-N
SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (15)
Quantity 
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (6)
  • (3)
  • (3)
  • (4)
  • (1)
  • (15)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)