Learn More
Gibberellic acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Gibberellic Acid |
| CAS | 77-06-5 |
| Analyte or Component Names | Gibberellic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 O6 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Gibb-3-ene-1,10-dicarboxylic acid, 2,4a,7-trihydroxy-1-methyl-8-methylene-, 1,4a-lactone, (1α,2β,4aα,4bβ,10β)-, Gibberellic acid (7CI,8CI), Gibberellin A3 (6CI), (+)-Gibberellic acid, (+)-Gibberellin A3, Activol, Arbostim, Berelex, Berelex L, Clemencuaje, GA, GA3, GibGro, Gibberellic acid GA3, Gibberellin GA3, Giberllon, Gibrescol, Gibreskol, Grocel GA3, Maxon, Maxon (plant growth regulator), NSC 14190, NSC 19450, Pro-Gibb, Pro-Gibb Plus, ProGib, Ralex, Release, Release LC, RyzUp, RyzUp SmartGrass, TU 64-3-103-75 |
| InChI Formula | InChI=1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1 |
| SMILES | C[C@]12[C@@H](O)C=C[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O |
| Show More |