CAS RN 76811-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Fexofenadinone, TRC
CAS: 76811-98-8 Molecular Formula: C32 H37 N O4 Molecular Weight (g/mol): 499.64 Synonym: 2-[4-[4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]butanoyl]phenyl]-2-methylpropanoic Acid,Fexofenadine Hydrochloride Imp. A (EP),Ketofexofenadine IUPAC Name: 2-[4-[4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butanoyl]phenyl]-2-methylpropanoic acid SMILES: CC(C)(C(=O)O)c1ccc(cc1)C(=O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
2
2-[4-[4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]butanoyl]phenyl]-2-methylpropanoic Acid (Ketofexofenadine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.