accessibility menu, dialog, popup

UserName

CAS RN 76-89-1

H3COOOH

CAS RN 76-89-1

IUPAC Name: methyl 2-hydroxy-2,2-diphenylacetate
Synonyms: methyl 2-hydroxy-2,2-diphenylacetate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: LJFIHTFNTGQZJL-UHFFFAOYSA-N
SMILES: COC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results