CAS RN 7550-35-8
Lithium bromide, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.84 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, 99+%, for analysis, anhydrous
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.84 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, anhydrous, 99% min
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, 99%, pure, anhydrous
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.84 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, anhydrous, 99.995% (metals basis)
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, anhydrous, 99.99% (metals basis)
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, ultra dry, 99.998% (metals basis)
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, ultra dry, 99.998% (metals basis)
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, ultra dry, 99.9% (metals basis)
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, anhydrous, 99% min
CAS: 7550-35-8 | BrLi
Lithium Bromide, Solstice
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: MFCD00011077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
Lithium bromide, 4M solution in THF, AcroSeal™
CAS: 7550-35-8 | BrLi | 86.84 g/mol
Lithium Bromide Anhydrous, Extra Pure, SLR, Fisher Chemical™
CAS: 7550-35-8 Molecular Formula: BrLi Molecular Weight (g/mol): 86.844 MDL Number: 11077 InChI Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M Synonym: lithium bromide,lithium monobromide,lithium bromide libr,lithiumbromide,libr,lithium 1+ ion bromide,lithium bromide, anhydrous,lithium bromide, ultra dry,bromolithium,lithium-bromide PubChem CID: 82050 ChEBI: CHEBI:63042 IUPAC Name: lithium;bromide SMILES: [Li+].[Br-]
- 1
- 2