missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bromide, Anhydrous, MP Biomedicals

Product Code. 11407514 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407514 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407514 Supplier MP Biomedicals Supplier No. 0520561925

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7550-35-8
Quantity 25 g
Molecular Formula BrLi
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES [Li+].[Br-]
IUPAC Name lithium;bromide
Molecular Weight (g/mol) 86.844
PubChem CID 82050
ChEBI CHEBI:63042
Formula Weight 86.84
Chemical Name or Material Lithium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.