accessibility menu, dialog, popup

UserName

ONNH2Br

CAS RN 754230-78-9

IUPAC Name: 3-(benzyloxy)-5-bromopyridin-2-amine
Synonyms: 3-(benzyloxy)-5-bromopyridin-2-amine
Molecular Weight (g/mol): 279.14
Molecular Formula: C12H11BrN2O
InChi Key: ZFNMAJBVWOHSSF-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results