accessibility menu, dialog, popup

UserName

H2NNH2H2N

CAS RN 74676-52-1

IUPAC Name: 1,5,9-triazacyclotridecane-1,5,9-triium
Synonyms: 1,5,9-triazacyclotridecane-1,5,9-triium
Molecular Weight (g/mol): 188.34
Molecular Formula: C10H26N3
InChi Key: UXABFDDIKPVEBR-UHFFFAOYSA-Q
SMILES: C1CC[NH2+]CCC[NH2+]CCC[NH2+]C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results