CAS RN 7440-25-7
Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 0.025mm (0.001in) dia, hard, 99.9% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum powder, -100 mesh, Puratronic™, 99.98% (metals basis), Nb 50ppm
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum powder, -325 mesh, 99.9% (metals basis), 99.6%
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod IUPAC Name: tantalum SMILES: [Ta]
Tantalum foil, 0.127mm (0.005in) thick, hard, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum rod, 2.8mm (0.11in) dia, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum gauze, 30 mesh woven from 0.076mm (0.003in) dia wire
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum gauze, 50 mesh woven from 0.076mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum foil, 0.127mm (0.005in) thick, annealed, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]
Tantalum slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)
CAS: 7440-25-7 Molecular Formula: Ta Molecular Weight (g/mol): 180.95 MDL Number: MFCD00011252 InChI Key: GUVRBAGPIYLISA-UHFFFAOYSA-N Synonym: tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod PubChem CID: 23956 ChEBI: CHEBI:33348 IUPAC Name: tantalum SMILES: [Ta]