CAS RN 7439-94-3
Lutetium pieces, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium pieces, distilled dendritic, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium powder, -40 mesh, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]
Lutetium Standard: Lu @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lutetium Standard: Lu @ 10 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lutetium Standard: Lu @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Rare Earth and 'Geo' Elements Standard: Ba, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Rb, Sc, Sm, Sr, Tb, Th, Tm, U, Y, Yb @ 100 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lutetium Internal Standard: Lu @ 10 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lutetium foil, 0.127mm (0.005in) thick, 99.9% (REO)
CAS: 7439-94-3 Molecular Formula: Lu Molecular Weight (g/mol): 174.97 MDL Number: MFCD00011098 InChI Key: OHSVLFRHMCKCQY-UHFFFAOYSA-N Synonym: lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces PubChem CID: 23929 ChEBI: CHEBI:33382 IUPAC Name: lutetium SMILES: [Lu]