CAS RN 74209-34-0
CAS RN 74209-34-0
IUPAC Name:
3-bromo-7-nitro-2H-indazole
Synonyms:
3-bromo-7-nitro-2H-indazole
Molecular Weight (g/mol):
242.03
Molecular Formula:
C7H4BrN3O2
InChi Key:
NFSTZPMYAZRZPC-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC2=C(Br)NN=C12
1
–
2
of
2
results
1
3-Bromo-7-nitroindazole, 98+%
CAS: 74209-34-0 Molecular Formula: C7H4BrN3O2 Molecular Weight (g/mol): 242.03 MDL Number: MFCD00159910 InChI Key: NFSTZPMYAZRZPC-UHFFFAOYSA-N Synonym: 3-bromo-7-nitroindazole,3-bromo-7-nitro-1h-indazole,1h-indazole,3-bromo-7-nitro,brni,3-bromo-7ni,3-br-7-nitroindazole,tocris-0735,lopac-b-2050,d0u6zt,d0x5qj PubChem CID: 1649 IUPAC Name: 3-bromo-7-nitro-2H-indazole SMILES: [O-][N+](=O)C1=CC=CC2=C(Br)NN=C12
2
Tocris Bioscience™ 3-Bromo-7-nitroindazole
CAS: 74209-34-0 Molecular Formula: C7H4BrN3O2