accessibility menu, dialog, popup

UserName

CAS RN 73096-42-1

IUPAC Name: 5-(2-bromophenyl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(2-bromophenyl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 224.04
Molecular Formula: C7H4BrN4
InChi Key: RONZSMDUKDEVRM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=N[N-]N=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results