accessibility menu, dialog, popup

UserName

CAS RN 73-24-5

IUPAC Name: 7H-purin-6-amine
Synonyms: adenine pedatisectine B 1H-purin-6-amine leuco-4 vitamin B4 adeninimine
Molecular Weight (g/mol): 135.13
Molecular Formula: C5H5N5
InChi Key: GFFGJBXGBJISGV-UHFFFAOYSA-N
SMILES: NC1=C2NC=NC2=NC=N1
Percent Purity 
  • (2)
  • (3)
  • (7)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (7)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)