CAS RN 72558-82-8
CAS RN 72558-82-8
IUPAC Name:
1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}pyridin-1-ium
Synonyms:
ceftazidime ceftazidimum ceftazidima [inn-spanish] SN 401 pentahydrate fortum fortaz tazidime in plastic container CAZ ceftazidima tazicef
Molecular Weight (g/mol):
546.57
Molecular Formula:
C22H22N6O7S2
InChi Key:
ORFOPKXBNMVMKC-DWVKKRMSSA-N
SMILES:
CC(C)(O\N=C(/C(=O)N[C@H]1[C@H]2SCC(C[N+]3=CC=CC=C3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1)C(O)=O
1
–
2
of
2
results
1
Melford Ceftazidime Hydrate
Ceftazidime is a third-generation cephalosporin antibiotic used in research settings. It is effective against a broad spectrum of bacteria, including Gram-negative and some Gram-positive bacteria.
2
Ceftazidime, MedChemExpress
MedChemExpress Ceftazidime (GR20263), an antibiotic, has a broad spectrum activity against Gram-positive and Gram-negative aerobic bacteria. Ceftazidime is also active against Enterobacteriaceae (including β-lactamase-positive strains) and is resistant to hydrolysis by most β-lactamases.