accessibility menu, dialog, popup

UserName

BONH3COCH3HOOH

CAS RN 723281-57-0

IUPAC Name: {3-[methoxy(methyl)carbamoyl]phenyl}boronic acid
Synonyms: 3-[methoxy(methyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 209.01
Molecular Formula: C9H12BNO4
InChi Key: IYZAWDLXZOCCSB-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results