accessibility menu, dialog, popup

UserName

CAS RN 72-57-1

NaNaNaNaCH3HNNONH2SOOOSOOONHNONH2SOOOSOOOCH3(Z)(Z)

CAS RN 72-57-1

IUPAC Name: tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: trypan blue
Molecular Weight (g/mol): 960.79
Molecular Formula: C34H24N6Na4O14S4
InChi Key: WMYVHJWZUUEZNE-ARWFNKCKSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
Color 
  • (3)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)