accessibility menu, dialog, popup

UserName

CAS RN 71888-97-6

CH3OSiNH2OH3COH3COSiO

CAS RN 71888-97-6

IUPAC Name: (3-aminopropyl)triethoxysilane; silanedione
Synonyms: aminopropyl triethoxysilane; silica
Molecular Weight (g/mol): 281.46
Molecular Formula: C9H23NO5Si2
InChi Key: JIBSXWZIQTVOKJ-UHFFFAOYSA-N
SMILES: O=[Si]=O.CCO[Si](CCCN)(OCC)OCC

 No Refinements