CAS RN 7179-49-9
CAS RN 7179-49-9
IUPAC Name:
hydrogen (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide hydrate chloride
Synonyms:
lincomycin
Molecular Weight (g/mol):
461.01
Molecular Formula:
C18H37ClN2O7S
InChi Key:
LFZGYTBWUHCAKF-DCNJEFSFSA-N
SMILES:
[H+].O.[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
1
–
3
of
3
results
2
Lincomycin hydrochloride monohydrate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
Lincomycin hydrochloride monohydrate, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality