missing translation for 'onlineSavingsMsg'
Learn More

Deltamethrin , MP Biomedicals™

Product Code. 11434152 Shop All MP Biomedicals Products
Change view
Click to view available options
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11434152 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434152 Supplier MP Biomedicals Supplier No. 0215889825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Deltamethrin, Powder

  • This is a Type II pyrethrin and a potent inhibitor of calcineurin (protein phosphatase 2B).
  • More recent study disputes its effectiveness as a calcineurin inhibitor.
  • May also have varied effects on ion channels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7179-49-9
Melting Point 98°C to 101°C
Color Off-white
Molecular Formula C18H37ClN2O7S
Quantity 25 mg
UN Number UN2811 II
Beilstein 6746312
Synonym (S)-α-cyano-3-phenoxybenzyl-(1R)-cis-3-(2,2-dibromvinyl)-2,2-dimethylcyclopropancarboxylate
Solubility Information Soluble in ethanol (25mg/mL) and DMSO (25mg/mL).
InChI Key LFZGYTBWUHCAKF-DCNJEFSFSA-N
SMILES [H+].O.[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name hydrogen (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide hydrate chloride
Molecular Weight (g/mol) 461.01
Vapor Pressure 0.002 mPa (25°C)
Physical Form Powder
Assay 0.983
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.