missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lincomycin hydrochloride monohydrate, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Lincomycin Hydrochloride Monohydrate |
| CAS | 7179-49-9 |
| Analyte or Component Names | Lincomycin Hydrochloride Monohydrate |
| Shipping Condition | +20°C |
| Molecular Formula | C18 H34 N2 O6 S . Cl H . H2 O |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, hydrochloride, hydrate (1:1:1), D-erythro-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, monohydrochloride, monohydrate, trans-α- (8CI), D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, monohydrochloride, monohydrate, (2S-trans)-, D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, monohydrochloride, monohydrate (9CI), Lincomycin hydrochloride hydrate, Lincomycin hydrochloride monohydrate, Ph Eur Clindamycin Phosphate Impurity A, Lincomycin hydrochloride monohydrate |
| InChI Formula | InChI=1S/C18H34N2O6S.ClH.H2O/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H;1H2/t9-,10-,11+,12-,13+,14-,15-,16-,18-;;/m1../s1 |
| SMILES | O.Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O |
| IUPAC Name | (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction