CAS RN 70916-89-1
IUPAC Name:
methyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate
Synonyms:
methyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol):
240.26
Molecular Formula:
C15H12O3
InChi Key:
URINKTWOCVLGGR-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
Methyl 4'-formylbiphenyl-4-carboxylate, 97%, Thermo Scientific™
CAS: 70916-89-1 Molecular Formula: C15H12O3 Molecular Weight (g/mol): 240.26 MDL Number: MFCD04039123 InChI Key: URINKTWOCVLGGR-UHFFFAOYSA-N Synonym: 4'-formylbiphenyl-4-carboxylic acid methyl ester,methyl 4'-formyl-1,1'-biphenyl-4-carboxylate,methyl 4-4-formylphenyl benzoate,methyl 4'-formyl 1,1'-biphenyl-4-carboxylate,acmc-209ogh,methyl 4'-formylbiphenyl-4-carboxylate,methyl-4'-formylbiphenyl-4-carboxylate,1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester,1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester PubChem CID: 4321323 IUPAC Name: methyl 4-(4-formylphenyl)benzoate SMILES: COC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1