accessibility menu, dialog, popup

UserName

HOO

CAS RN 708-06-5

IUPAC Name: 2-hydroxynaphthalene-1-carbaldehyde
Synonyms: 2-hydroxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: NTCCNERMXRIPTR-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1C=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)