Learn More
2-Hydroxy-1-naphthaldehyde, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 708-06-5 |
| Molecular Formula | C11H8O2 |
| MDL Number | MFCD00004005 |
| Quantity | 100 g |
| SMILES | O=CC1=C2C=CC=CC2=CC=C1O |
| Molecular Weight (g/mol) | 172.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Hydroxy-1-naphthaldehyde |