accessibility menu, dialog, popup

UserName

NH2OHNHSONHHOOOHO

CAS RN 70-18-8

IUPAC Name: 2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
Synonyms: reduced glutathione L-glutathione tathione glutathion
Molecular Weight (g/mol): 307.32
Molecular Formula: C10H17N3O6S
InChi Key: RWSXRVCMGQZWBV-UHFFFAOYNA-N
SMILES: NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (8)
  • (22)
Quantity 
  • (4)
  • (3)
  • (6)
  • (6)
  • (7)
  • (2)
  • (1)
Physical Form 
  • (8)
  • (12)
  • (6)
Percent Purity 
  • (14)
  • (3)
  • (5)
  • (2)
Grade 
  • (5)
  • (4)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (1)
  • (10)
  • (1)
  • (4)
  • (6)
  • (3)
  • (5)
  • (2)
  • (8)
  • (22)
  • (4)
  • (3)
  • (6)
  • (6)
  • (7)
  • (2)
  • (1)
  • (8)
  • (12)
  • (6)
  • (14)
  • (3)
  • (5)
  • (2)
  • (5)
  • (4)
  • (2)
  • (2)