CAS RN 688046-61-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Pyriofenone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Pyriofenone, TRC
CAS: 688046-61-9 Molecular Formula: C18H20ClNO5 Molecular Weight (g/mol): 365.81 Synonym: (5-Chloro-2-methoxy-4-methyl-3-pyridinyl)(2,3,4-trimethoxy-6-methylphenyl)methanone,3-(2,3,4-Trimethoxy-6-methylbenzoyl)-5-chloro-2-methoxy-4-methylpyridine,Chlazafenone,Clazafenone,IKF 309,Property Flowable,Pyriofenone IUPAC Name: (5-chloro-2-methoxy-4-methylpyridin-3-yl)-(2,3,4-trimethoxy-6-methylphenyl)methanone SMILES: COc1cc(C)c(C(=O)c2c(C)c(Cl)cnc2OC)c(OC)c1OC