missing translation for 'onlineSavingsMsg'
Learn More

Pyriofenone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30319497
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319497 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319497 Supplier Dr. Ehrenstorfer Supplier No. DREA16661500AL100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyriofenone
CAS 688046-61-9
Analyte or Component Names Pyriofenone
Shipping Condition +20°C
Molecular Formula C18H20ClNO5
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym (5-Chloro-2-methoxy-4-methyl-3-pyridinyl)(2,3,4-trimethoxy-6-methylphenyl)methanone, 3-(2,3,4-Trimethoxy-6-methylbenzoyl)-5-chloro-2-methoxy-4-methylpyridine, Chlazafenone, Clazafenone, IKF 309, Property Flowable, Pyriofenone
SMILES COc1cc(C)c(C(=O)c2c(C)c(Cl)cnc2OC)c(OC)c1OC
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 365.81
Quantity 1 mL
Formula Weight 365.103 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.