accessibility menu, dialog, popup

UserName

CAS RN 686770-61-6

CH3NSHNOSNONS

CAS RN 686770-61-6

IUPAC Name: N-(6-methyl-1,3-benzothiazol-2-yl)-2-({4-oxo-3-phenyl-3H,4H,6H,7H-thieno[3,2-d]pyrimidin-2-yl}sulfanyl)acetamide
Synonyms: N-(6-methyl-1,3-benzothiazol-2-yl)-2-({4-oxo-3-phenyl-6H,7H-thieno[3,2-d]pyrimidin-2-yl}sulfanyl)acetamide
Molecular Weight (g/mol): 466.59
Molecular Formula: C22H18N4O2S3
InChi Key: WRKPZSMRWPJJDH-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(NC(=O)CSC3=NC4=C(SCC4)C(=O)N3C3=CC=CC=C3)SC2=C1
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results