accessibility menu, dialog, popup

UserName

NCO

CAS RN 68664-23-3

IUPAC Name: (3-isocyanatopropyl)benzene
Synonyms: (3-isocyanatopropyl)benzene
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: WPABAKMRZCBXTO-UHFFFAOYSA-N
SMILES: O=C=NCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results