accessibility menu, dialog, popup

UserName

CAS RN 6815-00-5

IUPAC Name: (2-phenylethyl)thiourea
Synonyms: 2-phenylethylthiourea
Molecular Weight (g/mol): 180.27
Molecular Formula: C9H12N2S
InChi Key: OFVGPHQYOCKLLM-UHFFFAOYSA-N
SMILES: NC(=S)NCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results