accessibility menu, dialog, popup

UserName

HOONOONOHOHOOOHO

CAS RN 67-42-5

IUPAC Name: 3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Synonyms: EGTA ethylene glycol tetraacetic acid tetrasodium egta acido egtazico egta (chemical) Magnesium-EGTA egta, tetrasodium H4egta magnesium egta ethylene glycol bis(aminoethyl ether)tetraacetate
Molecular Weight (g/mol): 380.35
Molecular Formula: C14H24N2O10
InChi Key: DEFVIWRASFVYLL-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
Quantity 
  • (1)
  • (4)
  • (6)
  • (1)
  • (7)
  • (2)
  • (4)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (24)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (4)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (9)
Grade 
  • (2)
  • (4)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (10)
  • (2)
  • (3)
  • (6)
  • (1)
  • (3)
  • (5)
  • (1)
  • (4)
  • (6)
  • (1)
  • (7)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (24)
  • (3)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (9)
  • (2)
  • (4)
  • (1)
  • (2)