CAS RN 669091-00-3
IUPAC Name:
2-[bis(3,5-dimethylphenyl)phosphanyl]benzaldehyde
Synonyms:
2-[bis(3,5-dimethylphenyl)phosphanyl]benzaldehyde
Molecular Weight (g/mol):
346.41
Molecular Formula:
C23H23OP
InChi Key:
PQIAXRGPKDEYLW-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=CC=C1C=O
1
–
2
of
2
results
2
2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde, 97%, Thermo Scientific™
CAS: 669091-00-3 Molecular Formula: C23H23OP Molecular Weight (g/mol): 346.41 MDL Number: MFCD15143662 InChI Key: PQIAXRGPKDEYLW-UHFFFAOYSA-N Synonym: 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde,acmc-20ao6m,2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min PubChem CID: 71310677 IUPAC Name: 2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde SMILES: CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=CC=C1C=O