accessibility menu, dialog, popup

UserName

CAS RN 6656-72-0

FFFClH2NCl

CAS RN 6656-72-0

IUPAC Name: 2,6-dichloro-3-(trifluoromethyl)aniline
Synonyms: 2,6-dichloro-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 230.01
Molecular Formula: C7H4Cl2F3N
InChi Key: NKCROTQPQZXJOL-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC(=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results