CAS RN 66-23-9
CAS RN 66-23-9
IUPAC Name:
[2-(acetyloxy)ethyl]trimethylazanium bromide
Synonyms:
acetylcholine bromide tonocholin B bromoacetylcholine acetylcholine hydrobromide pragmoline acetylcholine bromhydrate
Molecular Weight (g/mol):
226.11
Molecular Formula:
C7H16BrNO2
InChi Key:
ZEHGKSPCAMLJDC-UHFFFAOYSA-M
SMILES:
[Br-].CC(=O)OCC[N+](C)(C)C
1
–
3
of
3
results
1
Acetylcholine Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Acetylcholine Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Acetylcholine Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.