Learn More
Acetylcholine Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | Acetylcholine Bromide |
| CAS | 66-23-9 |
| Molecular Formula | C7H16NO2.Br |
| Quantity | 5 g |
| Synonym | 2-(Acetyloxy)-N,N,N-trimethyl-ethanaminium Bromide, Choline Acetate (Ester) Bromide, (2-Acetyloxyethyl)trimethylammonium Bromide, Acetylcholine Bromhydrate, Acetylcholine Bromide, Acetylcholine Hydrobromide, N,N,N-Trimethyl-2-acetoxyethylammonium Bromide, Pragmoline, Tonocholin B, Trimethyl(2-acetoxyethyl)ammonium Bromide |
| InChI Formula | InChI=1S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1 |
| SMILES | [Br-].CC(=O)OCC[N+](C)(C)C |
| IUPAC Name | 2-acetyloxyethyl(trimethyl)azanium;bromide |
| Molecular Weight (g/mol) | 226.11 |
| Formula Weight | 225.0364 |