accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CONO

CAS RN 656239-38-2

IUPAC Name: 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Synonyms: 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Molecular Weight (g/mol): 317.19
Molecular Formula: C17H24BNO4
InChi Key: ZOPBEWUNCNLYSD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results