accessibility menu, dialog, popup

UserName

FFFONHONHOCl

CAS RN 64628-44-0

IUPAC Name: 1-(2-chlorobenzoyl)-3-[4-(trifluoromethoxy)phenyl]urea
Synonyms: triflumuron mascot 2-chloro-N-benzamide epa pesticide chemical code 118201 alsystine alsystin caswell no. 217C
Molecular Weight (g/mol): 358.7
Molecular Formula: C15H10ClF3N2O3
InChi Key: XAIPTRIXGHTTNT-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(NC(=O)NC(=O)C2=CC=CC=C2Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Concentration 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results