accessibility menu, dialog, popup

UserName

CAS RN 641571-10-0

CH3NNNHOCH3HNNNNFFF

CAS RN 641571-10-0

IUPAC Name: 4-methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}benzamide
Synonyms: nilotinib tasigna tasigna (novartis) nilotinibum
Molecular Weight (g/mol): 529.53
Molecular Formula: C28H22F3N7O
InChi Key: HHZIURLSWUIHRB-UHFFFAOYSA-N
SMILES: CC1=CN(C=N1)C1=CC(NC(=O)C2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)