accessibility menu, dialog, popup

UserName

ClH3CNCH3(S)(S)(S)OHOHCl(S)CH3HOO(S)OHOH2NOHOH(Z)

CAS RN 64-72-2

IUPAC Name: hydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Synonyms: chlortetracycline
Molecular Weight (g/mol): 515.34
Molecular Formula: C22H24Cl2N2O8
InChi Key: RRXOSDMYCQSQHB-MRYPCDLHSA-N
SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (15)
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Color 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (15)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)