Learn More
Chlorotetracycline hydrochloride, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Chlortetracycline Hydrochloride |
| CAS | 64-72-2 |
| Analyte or Component Names | Chlortetracycline Hydrochloride |
| Shipping Condition | +20°C |
| Molecular Formula | C22 H23 Cl N2 O8 . Cl H |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Chlortetracycline Hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, [4S-(4alpha,4aalpha,5aalpha,6beta,12aalpha)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, 7-Chlorotetracycline hydrochloride, 7-Chlorotetracycline monohydrochloride, Aureociclina, Aureocycline, Aureomycin hydrochloride, Aureomycin monohydrochloride, Aureovit 12C80, Aurofac 100, Biomycin hydrochloride, Chlortetracycline hydrochloride, Chlortetracyclinium chloride, Fermycin Soluble, Isphamycin, NSC 13252 |
| InChI Formula | InChI=1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7-,8-,15-,21-,22-;/m0./s1 |
| SMILES | Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O |
| IUPAC Name | (4S,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
| Show More |