accessibility menu, dialog, popup

UserName

CAS RN 63612-50-0

CH3H3CNHONFFFNOOO

CAS RN 63612-50-0

IUPAC Name: 5,5-dimethyl-3-[4-nitro-3-(trifluoromethyl)phenyl]imidazolidine-2,4-dione
Synonyms: nilutamide nilutamida anandron nilutamidum nilandrone nilandron nilutamide [usan:ban:inn]
Molecular Weight (g/mol): 317.22
Molecular Formula: C12H10F3N3O4
InChi Key: XWXYUMMDTVBTOU-UHFFFAOYSA-N
SMILES: CC1(C)NC(=O)N(C1=O)C1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results