Learn More
Nilutamide, MedChemExpress
Product Code. 16466286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16466286 Supplier Medchem Express Supplier No. HY13702
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
MedChemExpress Nilutamide (Nilandron) is a non-steroidal anti-androgen drug proposed in the treatment of metastatic prostatic carcinoma.
Nilutamide (Nilandron) is a non-steroidal anti-androgen drug proposed in the treatment of metastatic prostatic carcinoma.
Specifications
Chemical Name or Material Nilutamide
Physical Form Solid
CAS 63612-50-0
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H10F3N3O4
For Use With (Application) Cancer-programmed cell death
Synonym Nilandron RU 23908
Solubility Information DMSO : ≥ 250 mg/mL (788.10 mM)
SMILES O=C1N(C2=CC=C([N+]([O-])=O)C(C(F)(F)F)=C2)C(C(C)(C)N1)=O
Molecular Weight (g/mol) 317.22
Formula Weight 317.22
Percent Purity 98.07%
Grade Research