accessibility menu, dialog, popup

UserName

HBrNH2OO

CAS RN 6305-38-0

IUPAC Name: hydrogen 3-aminooxolan-2-one bromide
Synonyms: hydrogen HSLs bromide
Molecular Weight (g/mol): 182.02
Molecular Formula: C4H8BrNO2
InChi Key: MKLNTBLOABOJFZ-UHFFFAOYNA-N
SMILES: [H+].[Br-].NC1CCOC1=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)